Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Methoxy-2,5-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOB CHEM USA
Warning

58465-500MG 58465-1G 58465-25G 58465-5G
77330-296EA 300.76 USD InStock
77330-296 77354-362 77388-264 77378-470
(4-Methoxy-2,5-dimethylphenyl)(methyl)sulfane ≥97%
(4-Methoxy-2,5-dimethylphenyl)(methyl)sulfane
Precaución: for Research Use Only


Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD27929729
CAS Number: 84910-74-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty