Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-1-iodo-3-methylbenzene ≥97%

Supplier: AOB CHEM USA
Warning

58648-500MG 58648-1G 58648-5G 58648-25G
77330-324EA 126.69 USD InStock
77330-324 77344-158 77366-574 77378-530
2-Chloro-4-fluoro-1-iodo-3-methylbenzene ≥97%
2-Chloro-4-fluoro-1-iodo-3-methylbenzene
Precaución: for Research Use Only


Formula: C₇H₅ClFI
Storage Temperature: Ambient
MDL Number: MFCD28751660
CAS Number: 1056942-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty