Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Bromo-5-methylphenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOB CHEM USA
Warning

59391-500MG 59391-1G 59391-5G 59391-25G
77330-530EA 187.46 USD InStock
77330-530 77354-720 77366-742 77388-432
3-(2-Bromo-5-methylphenyl)-3-hydroxypropanoic acid ≥97%
3-(2-Bromo-5-methylphenyl)-3-hydroxypropanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD34759132
CAS Number: 2228194-96-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty