Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-isopropoxy-5-methoxybenzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

59422-500MG 59422-1G 59422-5G 59422-25G
77330-534EA 239.99 USD InStock
77330-534 77354-730 77366-744 77388-436
2-Fluoro-4-isopropoxy-5-methoxybenzoic acid ≥95%
2-Fluoro-4-isopropoxy-5-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD34759155
CAS Number: 1784406-31-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty