Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-isopropoxy-5-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

59822-500MG 59822-1G 59822-5G 59822-25G
77330-704EA 169.95 USD InStock
77330-704 77346-354 77366-918 77388-530
2-Fluoro-3-isopropoxy-5-methylbenzoic acid ≥97%
2-Fluoro-3-isopropoxy-5-methylbenzoic acid


Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34759427
CAS Number: 2755724-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty