Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-(4-bromophenoxy)-2-fluorobenzoate ≥95%

Supplier: AOB CHEM USA
Warning

60362-500MG 60362-1G 60362-25G 60362-5G
77330-864EA 300.76 USD InStock
77330-864 77355-078 77388-650 77379-154
Methyl 5-(4-bromophenoxy)-2-fluorobenzoate ≥95%
Methyl 5-(4-bromophenoxy)-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₁₄H₁₀BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34759706
CAS Number: 2764733-48-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty