Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-(4-bromophenoxy)-2-fluorobenzoate ≥95%

Supplier: AOB CHEM USA
Warning

60363-500MG 60363-1G 60363-5G
77330-866EA 313.12 USD InStock
77330-866 77355-080 77379-156
Ethyl 5-(4-bromophenoxy)-2-fluorobenzoate ≥95%
Ethyl 5-(4-bromophenoxy)-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₂BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34759707
CAS Number: 2764732-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty