Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)-4-ethoxybenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

60772-500MG 60772-1G 60772-5G 60772-25G
77331-022EA 172.01 USD InStock
77331-022 77346-422 77367-216 77388-750
3-(Benzyloxy)-4-ethoxybenzaldehyde ≥97%
3-(Benzyloxy)-4-ethoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₃
MW: 256.2964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00621686
CAS Number: 26691-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty