Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(tert-butyl)-2-ethoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

60922-500MG 60922-1G 60922-5G 60922-25G
77331-074EA 106.09 USD InStock
77331-074 77343-132 77367-260 77379-368
3-Bromo-5-(tert-butyl)-2-ethoxybenzoic acid ≥97%
3-Bromo-5-(tert-butyl)-2-ethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₇BrO₃
MW: 301.1762 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760034
CAS Number: 2217024-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty