Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-3-(hydroxymethyl)phenol ≥97%

Supplier: AOB CHEM USA
Warning

61184-500MG 61184-1G 61184-5G
77331-148EA 334.75 USD InStock
77331-148 77355-374 77379-450
2-Chloro-4-fluoro-3-(hydroxymethyl)phenol ≥97%
2-Chloro-4-fluoro-3-(hydroxymethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28553799
CAS Number: 1782418-39-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty