Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

61279-500MG 61279-1G 61279-5G 61279-25G
77331-182EA 136.99 USD InStock
77331-182 77344-604 77367-358 77388-876
5-Fluoro-2,4-dimethoxybenzoic acid ≥97%
5-Fluoro-2,4-dimethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₉H₉FO₄
MW: 200.1637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24481271
CAS Number: 430459-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty