Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,6-dichloro-4-(trifluoromethoxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

61507-500MG 61507-1G 61507-5G 61507-25G
77331-268EA 204.97 USD InStock
77331-268 77355-538 77367-408 77388-926
Ethyl 2,6-dichloro-4-(trifluoromethoxy)benzoate ≥97%
Ethyl 2,6-dichloro-4-(trifluoromethoxy)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₇Cl₂F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34760381
CAS Number: 2703252-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty