Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Bromo-6-iodo-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOB CHEM USA
Warning

62328-500MG 62328-1G 62328-5G 62328-25G
77331-612EA 178.19 USD InStock
77331-612 77346-668 77367-740 77389-168
(2-Bromo-6-iodo-4-methylphenyl)(methyl)sulfane ≥97%
(2-Bromo-6-iodo-4-methylphenyl)(methyl)sulfane


Formula: C₈H₈BrIS
Storage Temperature: Ambient
MDL Number: MFCD34760804
CAS Number: 2707848-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty