Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-fluoro-5-methoxy-3-methylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

62466-500MG 62466-1G 62466-5G 62466-25G
77331-662EA 201.88 USD InStock
77331-662 77355-890 77367-794 77389-194
Ethyl 2-fluoro-5-methoxy-3-methylbenzoate ≥97%
Ethyl 2-fluoro-5-methoxy-3-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃FO₃
MW: 212.2175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16038789
CAS Number: 1260765-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty