Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-formyl-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

62702-500MG 62702-1G 62702-5G 62702-25G
77331-746EA 107.12 USD InStock
77331-746 77343-134 77367-884 77380-036
2-Fluoro-3-formyl-5-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-formyl-5-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₄F₄O₃
MW: 236.1198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760992
CAS Number: 2385457-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty