Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-4',5-difluoro-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOB CHEM USA
Warning

64362-500MG 64362-1G 64362-25G 64362-5G
77332-170EA 296.64 USD InStock
77332-170 77356-540 77389-558 77380-478
4-Bromo-4',5-difluoro-2-methyl-1,1'-biphenyl ≥97%
4-Bromo-4',5-difluoro-2-methyl-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₁₃H₉BrF₂
Storage Temperature: Ambient
MDL Number: MFCD34761737
CAS Number: 2807454-66-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty