Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-(2-methoxyethoxy)-5-methylpyridine ≥97%

Supplier: AOB CHEM USA
Warning

64972-500MG 64972-1G 64972-5G 64972-25G
77332-374EA 136.99 USD InStock
77332-374 77344-562 77368-440 77389-700
3-Bromo-2-(2-methoxyethoxy)-5-methylpyridine ≥97%
3-Bromo-2-(2-methoxyethoxy)-5-methylpyridine
Precaución: for Research Use Only


Formula: C₉H₁₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD29114189
CAS Number: 1289009-92-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty