Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-5-hydroxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

65285-500MG 65285-1G 65285-5G
77337-766EA 440.84 USD InStock
77337-766 77356-808 77380-752
2-Chloro-4-fluoro-5-hydroxybenzoic acid ≥97%
2-Chloro-4-fluoro-5-hydroxybenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD32696816
CAS Number: 91659-15-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty