Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Fluoro-4-methyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOB CHEM USA
Warning

66064-500MG 66064-1G 66064-5G 66064-25G
77332-734EA 184.37 USD InStock
77332-734 77347-296 77368-778 77389-964
3'-Fluoro-4-methyl-[1,1'-biphenyl]-3-amine ≥97%
3'-Fluoro-4-methyl-[1,1'-biphenyl]-3-amine
Precaución: for Research Use Only


Formula: C₁₃H₁₂FN
Storage Temperature: Ambient
MDL Number: MFCD12824104
CAS Number: 1183439-66-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty