Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Tribromo-1-ethyl-1H-imidazole ≥97%

Supplier: AOB CHEM USA
Warning

66461-500MG 66461-1G 66461-5G 66461-25G
77332-798EA 95.79 USD InStock
77332-798 77342-584 77368-846 77380-978
2,4,5-Tribromo-1-ethyl-1H-imidazole ≥97%
2,4,5-Tribromo-1-ethyl-1H-imidazole
Precaución: for Research Use Only


Formula: C₅H₅Br₃N₂
Storage Temperature: Ambient
MDL Number: MFCD11868105
CAS Number: 31250-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty