Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-6-methoxy-2-methylbenzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

67990-500MG 67990-1G 67990-5G 67990-25G
77333-098EA 279.13 USD InStock
77333-098 77357-302 77369-118 77390-228
3-Bromo-6-methoxy-2-methylbenzoic acid ≥95%
3-Bromo-6-methoxy-2-methylbenzoic acid


Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD01846084
CAS Number: 220901-25-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty