Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxy-5-fluoro-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

68004-500MG 68004-1G 68004-5G
77339-468EA 592.25 USD InStock
77339-468 77357-312 77381-216
4-Ethoxy-5-fluoro-2-(trifluoromethyl)benzoic acid ≥97%
4-Ethoxy-5-fluoro-2-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763106
CAS Number: 2703616-06-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty