Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-3-(trimethylsilyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

68873-500MG 68873-1G 68873-25G 68873-5G
77333-322EA 298.7 USD InStock
77333-322 77357-500 77390-372 77381-450
2,4-Difluoro-3-(trimethylsilyl)benzoic acid ≥97%
2,4-Difluoro-3-(trimethylsilyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂F₂O₂Si
Storage Temperature: Ambient
MDL Number: MFCD30178436
CAS Number: 872545-54-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty