Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-6-propoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

71388-500MG 71388-1G 71388-5G 71388-25G
77333-602EA 204.97 USD InStock
77333-602 77357-738 77369-614 77390-564
2,3-Difluoro-6-propoxybenzoic acid ≥97%
2,3-Difluoro-6-propoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34764114
CAS Number: 2918870-57-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty