Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Ethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOB CHEM USA
Warning

71711-500MG 71711-1G 71711-5G 71711-25G
77333-664EA 210.12 USD InStock
77333-664 77357-818 77369-672 77390-620
1-(4-Ethylphenyl)-2,2-difluoroethanone ≥95%
1-(4-Ethylphenyl)-2,2-difluoroethanone
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₂O
MW: 184.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618094
CAS Number: 1352216-96-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty