Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-5-propoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

72263-500MG 72263-1G 72263-5G 72263-25G
77333-690EA 219.39 USD InStock
77333-690 77357-872 77369-698 77390-646
3-Methyl-5-propoxybenzoic acid ≥97%
3-Methyl-5-propoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₃
MW: 194.2270 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28247301
CAS Number: 1823833-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty