Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-6-chlorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

72728-500MG 72728-1G 72728-5G 72728-25G
77333-794EA 235.87 USD InStock
77333-794 77357-966 77369-810 77390-718
2,4-Dibromo-6-chlorobenzoic acid ≥97%
2,4-Dibromo-6-chlorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Br₂ClO₂
Storage Temperature: Ambient
MDL Number: MFCD30178561
CAS Number: 65410-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty