Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Propoxy-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

73185-500MG 73185-1G 73185-25G 73185-5G
77333-892EA 305.91 USD InStock
77333-892 77358-048 77390-784 77381-974
3-Propoxy-5-(trifluoromethoxy)benzoic acid ≥97%
3-Propoxy-5-(trifluoromethoxy)benzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁F₃O₄
MW: 264.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764793
CAS Number: 2807447-90-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty