Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-isobutoxy-3,5-dimethylbenzene ≥97%

Supplier: AOB CHEM USA
Warning

73541-500MG 73541-1G 73541-5G 73541-25G
77333-924EA 129.78 USD InStock
77333-924 77344-316 77369-936 77382-000
1-Bromo-2-isobutoxy-3,5-dimethylbenzene ≥97%
1-Bromo-2-isobutoxy-3,5-dimethylbenzene
Precaución: for Research Use Only


Formula: C₁₂H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD16330831
CAS Number: 1226307-72-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty