Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyl-4-(benzyloxy)benzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

73783-500MG 73783-1G 73783-5G
77338-860EA 526.33 USD InStock
77338-860 77358-110 77382-024
3-Benzyl-4-(benzyloxy)benzaldehyde ≥97%
3-Benzyl-4-(benzyloxy)benzaldehyde
Precaución: for Research Use Only


Formula: C₂₁H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34765150
CAS Number: 343870-59-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty