Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluoro-3-(trimethylsilyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

73930-500MG 73930-1G 73930-5G 73930-25G
77333-946EA 190.55 USD InStock
77333-946 77358-126 77369-954 77390-824
5-Chloro-2-fluoro-3-(trimethylsilyl)benzoic acid ≥97%
5-Chloro-2-fluoro-3-(trimethylsilyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClFO₂Si
Storage Temperature: Ambient
CAS Number: 2921860-53-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty