Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-2,4,6-trifluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

74334-500MG 74334-1G 74334-5G 74334-25G
77334-000EA 99.91 USD InStock
77334-000 77342-746 77370-000 77382-076
3-Ethoxy-2,4,6-trifluorobenzoic acid ≥97%
3-Ethoxy-2,4,6-trifluorobenzoic acid
Precaución: for Research Use Only


Formula: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34765391
CAS Number: 2807448-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty