Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-bromo-5-fluoro-4-iodobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

76114-500MG 76114-1G 76114-5G
77339-104EA 550.02 USD InStock
77339-104 77358-386 77382-284
Ethyl 2-bromo-5-fluoro-4-iodobenzoate ≥97%
Ethyl 2-bromo-5-fluoro-4-iodobenzoate
Precaución: for Research Use Only


Formula: C₉H₇BrFIO₂
MW: 372.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766279
CAS Number: 2385284-61-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty