Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-6-fluoro-2-(trifluoromethyl)-5-(trimethylsilyl)benzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

76419-500MG 76419-1G 76419-25G 76419-5G
77334-234EA 297.67 USD InStock
77334-234 77358-420 77391-038 77382-310
3-Bromo-6-fluoro-2-(trifluoromethyl)-5-(trimethylsilyl)benzoic acid ≥95%
3-Bromo-6-fluoro-2-(trifluoromethyl)-5-(trimethylsilyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁BrF₄O₂Si
Storage Temperature: Ambient
MDL Number: MFCD34766506
CAS Number: 2918847-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty