Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylbenzo[b]thiophene-3-carboxylic acid ≥95%

Supplier: AOB CHEM USA
Warning

76783-500MG 76783-1G 76783-5G 76783-25G
77334-258EA 119.48 USD InStock
77334-258 77343-650 77370-268 77382-334
2-Methylbenzo[b]thiophene-3-carboxylic acid ≥95%
2-Methylbenzo[b]thiophene-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD11856810
CAS Number: 17375-82-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty