Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2,5-dimethoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

77142-500MG 77142-1G 77142-5G
77338-436EA 487.19 USD InStock
77338-436 77358-498 77382-388
4-Fluoro-2,5-dimethoxybenzoic acid ≥97%
4-Fluoro-2,5-dimethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₉H₉FO₄
Storage Temperature: Ambient
MDL Number: MFCD28640275
CAS Number: 1553751-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty