Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,6-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

77373-500MG 77373-1G 77373-5G 77373-25G
77334-314EA 176.13 USD InStock
77334-314 77346-564 77370-316 77391-090
Ethyl 2,6-bis(trifluoromethyl)benzoate ≥97%
Ethyl 2,6-bis(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₁H₈F₆O₂
Storage Temperature: Ambient
MDL Number: MFCD12025039
CAS Number: 38570-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty