Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4'-methoxy-5-methyl-1,1'-biphenyl ≥97%

Supplier: AOB CHEM USA
Warning

78430-500MG 78430-1G 78430-5G 78430-25G
77334-406EA 114.33 USD InStock
77334-406 77343-376 77370-388 77382-504
2-Chloro-4'-methoxy-5-methyl-1,1'-biphenyl ≥97%
2-Chloro-4'-methoxy-5-methyl-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₁₄H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD34767514
CAS Number: 1357162-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty