Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Chloro-5'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

78431-500MG 78431-1G 78431-5G 78431-25G
77334-408EA 264.71 USD InStock
77334-408 77358-626 77370-390 77391-152
2'-Chloro-5'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Chloro-5'-methyl-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₄H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD34767515
CAS Number: 1999982-93-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty