Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-3-iodobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

80569-500MG 80569-1G 80569-5G
77340-378EA 978.5 USD InStock
77340-378 77358-754 77382-676
2,5-Dichloro-3-iodobenzoic acid ≥97%
2,5-Dichloro-3-iodobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Cl₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD07780706
CAS Number: 42860-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty