Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,3-Difluoro-4-methylphenyl)trimethylsilane ≥95%

Supplier: AOB CHEM USA
Warning

80626-500MG 80626-1G 80626-5G 80626-25G
77334-552EA 243.08 USD InStock
77334-552 77358-756 77370-550 77391-246
(2,3-Difluoro-4-methylphenyl)trimethylsilane ≥95%
(2,3-Difluoro-4-methylphenyl)trimethylsilane


Formula: C₁₀H₁₄F₂Si
Storage Temperature: Ambient
MDL Number: MFCD34768427
CAS Number: 1862935-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty