Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-ethylbenzoylformate ≥97%

Supplier: AOB CHEM USA
Warning

82409-500MG 82409-1G 82409-5G 82409-25G
77334-644EA 131.84 USD InStock
77334-644 77344-380 77370-666 77391-318
Ethyl-4-ethylbenzoylformate ≥97%
Ethyl 4-ethylbenzoylformate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD06201713
CAS Number: 62936-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty