Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(4-isobutylphenyl)-2-oxoacetate ≥95%

Supplier: AOB CHEM USA
Warning

82559-500MG 82559-1G 82559-5G 82559-25G
77334-702EA 126.69 USD InStock
77334-702 77344-222 77370-726 77382-818
Ethyl 2-(4-isobutylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-isobutylphenyl)-2-oxoacetate
Precaución: for Research Use Only


Formula: C₁₄H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD11113527
CAS Number: 60473-28-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty