Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-dimethoxyphenyl acetate ≥97%

Supplier: AOB CHEM USA
Warning

85018-500MG 85018-1G 85018-5G
77334-792EA 348.14 USD InStock
77334-792 77358-950 77382-916
3,4-dimethoxyphenyl acetate ≥97%
3,4-dimethoxyphenyl acetate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD16877752
CAS Number: 7203-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty