Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-ethylthiophene ≥97%

Supplier: AOB CHEM USA
Warning

85322-500MG 85322-1G 85322-5G 85322-25G
77334-816EA 143.17 USD InStock
77334-816 77344-992 77370-852 77391-432
4-Bromo-2-ethylthiophene ≥97%
4-Bromo-2-ethylthiophene
Precaución: for Research Use Only


Formula: C₆H₇BrS
Storage Temperature: Ambient
MDL Number: MFCD15976358
CAS Number: 83663-40-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty