Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-nitrophenyl acetate ≥97%

Supplier: AOB CHEM USA
Warning

86103-500MG 86103-1G 86103-5G 86103-25G
77334-876EA 89.61 USD InStock
77334-876 77342-352 77370-912 77382-992
2,4-Dichloro-5-nitrophenyl acetate ≥97%
2,4-Dichloro-5-nitrophenyl acetate
Precaución: for Research Use Only


Formula: C₈H₅Cl₂NO₄
Storage Temperature: Ambient
CAS Number: 123871-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty