Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl(4-(trifluoromethoxy)phenyl)silane ≥97%

Supplier: AOB CHEM USA
Warning

86152-500MG 86152-1G 86152-5G 86152-25G
77334-894EA 243.08 USD InStock
77334-894 77359-042 77370-930 77391-492
Trimethyl(4-(trifluoromethoxy)phenyl)silane ≥97%
Trimethyl(4-(trifluoromethoxy)phenyl)silane
Precaución: for Research Use Only


Formula: C₁₀H₁₃F₃OSi
MW: 234.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370325
CAS Number: 1443303-48-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty