Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

87107-500MG 87107-1G 87107-5G
77336-018EA 373.89 USD InStock
77336-018 77359-094 77383-030
3-Ethoxy-2,4-dimethoxybenzoic acid ≥97%
3-Ethoxy-2,4-dimethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00597855
CAS Number: 33257-20-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty