Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isobutoxy-2-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

88436-500MG 88436-1G 88436-5G 88436-25G
77335-000EA 201.88 USD InStock
77335-000 77359-132 77371-034 77391-566
4-Isobutoxy-2-methylbenzoic acid ≥97%
4-Isobutoxy-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19309240
CAS Number: 175153-57-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty