Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-diethoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

91759-500MG 91759-1G 91759-5G
77339-720EA 616.97 USD InStock
77339-720 77359-224 77383-188
2,3-diethoxybenzoic acid ≥97%
2,3-diethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD11652697
CAS Number: 37860-70-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty